Learn More
Cayman Chemical Galactosylceramide (non-hydroxy) (bovine), Purity: ≥98%, CAS: 536-13-0, 10 mg
SDP
Catalog No. 502932469
Encompass_Preferred
A mixture of isolated bovine non-hydroxy galactosylceramides; metabolic precursors to sulfatide; found primarily in nerve tissues; involved in cell agglutination, cellular signaling in glycosynapses, cellular development, and activation of T cells; accumulate in globoid cells in the brain of patients with Krabbe disease
Catalog No. 50-293-2469 Supplier Cayman Chemical Supplier No. 2483510 MG
A mixture of isolated bovine non-hydroxy galactosylceramides; metabolic precursors to sulfatide; found primarily in nerve tissues; involved in cell agglutination, cellular signaling in glycosynapses, cellular development, and activation of T cells; accumulate in globoid cells in the brain of patients with Krabbe disease