Learn More
Cayman Chemical GalactosylcRamIdShydroxy 10mg
SDP
Catalog No. 502933512
Encompass_Preferred
A sphingolipid; metabolic precursors to sulfatides; found primarily in nerve tissues; involved in cell agglutination, cellular signaling in glycosynapses, cellular development, and activation of T cells; accumulate in globoid cells in the brain of patients with Krabbe disease
Catalog No. 50-293-3512 Supplier Cayman Chemical Supplier No. 2483610 MG
A sphingolipid; metabolic precursors to sulfatides; found primarily in nerve tissues; involved in cell agglutination, cellular signaling in glycosynapses, cellular development, and activation of T cells; accumulate in globoid cells in the brain of patients with Krabbe disease