Learn More
Medchemexpress LLC HY-P1578A 1mg Medchemexpress, Galanin (1-16), mouse, porcine, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1578A1MG
Encompass_Preferred
Medchemexpress, HY-P1578A 1mg Galanin (1-16), mouse, porcine, rat (TFA) CAS: Galanin (1-16), mouse, porcine, rat (TFA) is an agonist of the hippocampal galanin receptor , with a K d of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4229