Learn More
TARGETMOL CHEMICALS INC Galanin 1-16 mouse porcine rat
SDP
Catalog No. 502877190
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Galanin (1-16), mouse, porcine, rat, is a hippocampal galanin receptor agonist (Kd = 3 nM) exhibiting high biological activity on locus coeruleus neurons.
Catalog No. 50-287-7190 Supplier TARGETMOL CHEMICALS INC Supplier No. TP13801MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Galanin (1-16), mouse, porcine, rat, is a hippocampal galanin receptor agonist (Kd = 3 nM) exhibiting high biological activity on locus coeruleus neurons.