Learn More
Medchemexpress LLC Galanin (1-30), human | 119418-04-1 | 99.9% | 3157.46 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP1127500UG
Encompass_Preferred
Galanin (1-30), human is a 30-amino acid neuropeptide that acts as an agonist of GalR1 and GalR2 receptors, with Kis of both 1 nM.
  • Agonist of GalR1 and GalR2 receptors, with Kis of both 1 nM.
  • Displaces 125I-labeled rat galanin with a Kd of 0.5 nM.
  • Causes contractions of isolated longitudinal rat fundus strips, with an ED50 of 13.8 ± 1.6 nM.
Catalog No. 50-004-24600