Learn More
Santa Cruz Biotechnology GALANIN ANTIBODY(H-11)200UG/ML
Supplier:  Santa Cruz Biotechnology SC166431
Encompass
Galanin Antibody (H-11)200 µg/mlGalanin Antibody (H-11) is a mouse monoclonal IgG1 κ Galanin antibody, cited in 7 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 104-123 near the C-terminus of Galanin of human originGalanin Antibody (H-11) is recommended for detection of Galanin precursor and Galanin message-associated peptide (GMAP) of human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC2226078