Learn More
Bachem Galanin (human) Acetate
SDP
Supplier:  Bachem 4095896.1000
Encompass_Preferred
Galanin (human) Acetate, H-Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Val-Gly-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-Asn-Gly-Leu-Thr-Ser-OH, 119418-04-1, C139H210N42O43, Mr 3157.45, 1mg. Galanin, human. Galanin has been shown to inhibit acetylcholine release, glutamate release and carbachol-stimulated phosphatidylinositol hydrolysis in the hippocampus. The peptide hormone is also linked to behavioral and cognitive deficits in Alzheimer's disease.
Catalog No. 50-194-6644