Learn More
Enzo Life Sciences Galanin (human) (1mg). CAS: 119418-04-1
SDP
Supplier:  Enzo Life Sciences ALX157003P001
Encompass_Preferred
Neuropeptide widely expressed in the central and peripheral nervous systems and in the endocrine system. Has been shown to regulate numerous physiological and pathological processes through interactions with three G-protein-coupled receptors, GalR1 through GalR3. Formula: C139H210N42O43. MW: 3157.5. Purity: ≥98%. Solubility: Soluble in water, 0.1% TFA or acetic acid solution. Long Term Storage: -20°C.
Catalog No. 50-200-7819