Learn More
CPC Scientific H-Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Val-Gly-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-Asn-Gly-Leu-Thr-Ser-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GALA001A
Encompass
Human galanin, originally been isolated from colon and pituitary, that activates the rat galanin receptor and inhibits acetylcholine release, glutamate release and carbachol-stimulated phosphatidylinositol hydrolysis in the hippocampus. Recent studies indicated that galanin is also linked to behavioral and cognitive deficits in Alzheimer's disease. ONE-LETTER SEQUENCE: GWTLNSAGYLLGPHAVGNHRSFSDKNGLTSMOLECULAR FORMULA: C139H210N42O43MOLECULAR WEIGHT:3157.45STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [119418-04-1], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: W.E. Schmidt et al., PNAS, 88, 11435 (1991)
Catalog No. 50-210-9304