Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Galanin protein, human (N-His, C-Myc) | >90.0% | 50UG
SDP

Catalog No. 5000176558
Encompass_Preferred

Galanin protein, human (N-His, C-Myc) is a recombinant human-derived galanin fragment (residues G44-S123) provided for research applications. The protein carries an N-terminal 10xHis tag and a C-terminal c-Myc tag, is expressed in Escherichia coli, and is supplied with batch-specific documentation for characterization and quality control.

  • Recombinant human galanin fragment G44-S123.
  • N-terminal 10xHis and C-terminal c-Myc tags.
  • Expressed in Escherichia coli for efficient production.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Predicted molecular weight 16.4 kDa; observed ~18 kDa on SDS-PAGE.
  • Batch-specific data sheet, SDS, and COA available.

Catalog No. 50-001-76558 Supplier Medchemexpress LLC Supplier No. HYP70056450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Galanin protein, human (N-His, C-Myc) is a recombinant human-derived galanin fragment (residues G44-S123) provided for research applications. The protein carries an N-terminal 10xHis tag and a C-terminal c-Myc tag, is expressed in Escherichia coli, and is supplied with batch-specific documentation for characterization and quality control.

  • Recombinant human galanin fragment G44-S123.
  • N-terminal 10xHis and C-terminal c-Myc tags.
  • Expressed in Escherichia coli for efficient production.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Predicted molecular weight 16.4 kDa; observed ~18 kDa on SDS-PAGE.
  • Batch-specific data sheet, SDS, and COA available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.