Learn More
Echelon Biosciences Research Labs Galanin, porcine - 0.5 mg, CAS#: 88813-36-9
SDP
Supplier:  Echelon Biosciences Research Labs 471250.5MG
Encompass_Preferred
Galanin, porcine - 0.5mg , Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile-Asp-Asn-His-Arg-Ser-Phe-His-Asp-Lys-Tyr-Gly-Leu-Ala-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8482