Learn More
CPC Scientific H-Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile-Asp-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-His-Gly-Leu-Thr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GALA003A
Encompass
ONE-LETTER SEQUENCE: GWTLNSAGYLLGPHAIDNHRSFSDKHGLT-NH2MOLECULAR FORMULA: C141H211N43O41MOLECULAR WEIGHT:3164.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [114547-31-8], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: M.E. Vrontakis et al., JBC, 262, 16755 (1987); L.M. Kaplan et al., PNAS, 85, 1065 (1988)
Catalog No. 50-210-9308