Learn More
Medchemexpress LLC HY-P1840 1mg Medchemexpress, Galanin Receptor Ligand M35 CAS:142846-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP18401MG
Encompass_Preferred
Medchemexpress, HY-P1840 1mg Galanin Receptor Ligand M35 CAS:142846-71-7 Galanin Receptor Ligand M35 is a high-affinity ligand and antagonist of galanin receptor (Kd=0.1 nM). Galanin Receptor Ligand M35 exerts a Ki values of 0.11 and 2.0 nM for human galanin receptor type 1 and 2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3074