Learn More
Abcam Galanthamine, cholinesterase inhibitor, 500MG
Catalog No. 502683362
Encompass_Preferred
MW 287.35 Da. Potent selective acetylcholinesterase inhibitor (IC₅₀ = 410 nM). Greater than 50-fold selective versus butyrylcholinesterase. Inhibits RBC cholinesterase activity 10-fold more potently than that of human brain tissue. Reduces apoptotic effect of thapsigargin in SH-SY5Y cells. Non-competitive nicotinic receptor aganist in rat PC12 cells. Neuroprotective following ischemia-reperfusion injury in animal models.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3362 Supplier Abcam Supplier No. AB269700500MG
MW 287.35 Da. Potent selective acetylcholinesterase inhibitor (IC₅₀ = 410 nM). Greater than 50-fold selective versus butyrylcholinesterase. Inhibits RBC cholinesterase activity 10-fold more potently than that of human brain tissue. Reduces apoptotic effect of thapsigargin in SH-SY5Y cells. Non-competitive nicotinic receptor aganist in rat PC12 cells. Neuroprotective following ischemia-reperfusion injury in animal models.
The product is subject to the following: Abcam Restricted Use Statement