Learn More
Medchemexpress LLC Galanthamine-d6 | 1128109-00-1 | 99.5% | C17H15D6NO3 | 5 MG
SDP
Catalog No. 5000434684
Encompass_Preferred
Galanthamine-d6 (Galantamine-d6) is a deuterium-labeled form of Galanthamine, a potent acetylcholinesterase (AChE) inhibitor with an IC50 of 500 nM. This compound is valuable as a tracer and an internal standard for quantitative analysis. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making this labeled compound useful in drug development research.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent acetylcholinesterase (AChE) inhibitor
  • Influences pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34684 Supplier Medchemexpress LLC Supplier No. HY76299S5MG
Galanthamine-d6 (Galantamine-d6) is a deuterium-labeled form of Galanthamine, a potent acetylcholinesterase (AChE) inhibitor with an IC50 of 500 nM. This compound is valuable as a tracer and an internal standard for quantitative analysis. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making this labeled compound useful in drug development research.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent acetylcholinesterase (AChE) inhibitor
  • Influences pharmacokinetic and metabolic profiles of drugs