Learn More
Medchemexpress LLC Galanthamine-d6 | 1128109-00-1 | MFCD28138526 | 99.5% | 293.39 | C17H15D6NO3 | 10 MG
SDP
Catalog No. 5000298663
Encompass_Preferred
Galanthamine-d6 is the deuterium-labeled analogue of galanthamine, a potent acetylcholinesterase (AChE) inhibitor. The compound is intended for research use and serves as an analytical standard or internal standard for quantitative analysis techniques such as NMR, GC-MS, and LC-MS.
  • Deuterium-d6 labeled galanthamine for isotope tracing and analytical standard use.
  • High isotopic and chemical purity (reported ~99.5%).
  • Appropriate as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small sizes for analytical workflows (10 mg available).
  • Solid, white to yellow appearance; powder storage recommended at -20°C for long-term stability; solvent samples require lower temperatures.
Catalog No. 50-002-98663 Supplier Medchemexpress LLC Supplier No. HY76299S10MG
Galanthamine-d6 is the deuterium-labeled analogue of galanthamine, a potent acetylcholinesterase (AChE) inhibitor. The compound is intended for research use and serves as an analytical standard or internal standard for quantitative analysis techniques such as NMR, GC-MS, and LC-MS.
  • Deuterium-d6 labeled galanthamine for isotope tracing and analytical standard use.
  • High isotopic and chemical purity (reported ~99.5%).
  • Appropriate as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small sizes for analytical workflows (10 mg available).
  • Solid, white to yellow appearance; powder storage recommended at -20°C for long-term stability; solvent samples require lower temperatures.