Learn More
Medchemexpress LLC Galanthamine hydrobromide | 1953-04-4 | MFCD00067672 | 99.9% | C17H22BrNO3 | 100 MG
Supplier: Medchemexpress LLC HYA0009100MG
Galanthamine hydrobromide is a selective, reversible, and competitive alkaloid acetylcholinesterase (AChE) inhibitor with an IC50 of 0.35 μM. It also acts as a potent allosteric potentiating ligand (APL) for human α3β4, α4β2, and α6β4 nicotinic receptors (nAChRs). This compound is developed for research related to Alzheimer's disease (AD).
- Selective, reversible, competitive AChE inhibitor
- Potent allosteric potentiating ligand for human nicotinic receptors
- Reduces cognitive deficits in APP23 mice
- Protects against Aβ toxicity in SH-SY5Y cells