Learn More
TARGETMOL CHEMICALS INC Galanthamine hydrobromide
SDP
Catalog No. 502880562
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Galanthamine hydrobromide (Galantamine hydrobromide), a long-acting and centrally active AChE inhibitor (IC50410 nM), is an allosteric potentiator at neuronal nicotinic ACh receptors. Purity 99.19%
Catalog No. 50-288-0562 Supplier TARGETMOL CHEMICALS INC Supplier No. T0086500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Galanthamine hydrobromide (Galantamine hydrobromide), a long-acting and centrally active AChE inhibitor (IC50410 nM), is an allosteric potentiator at neuronal nicotinic ACh receptors. Purity 99.19%