Learn More
Medchemexpress LLC Galanthamine hydrobromide (Galantamine hydrobromide) | 1953-04-4 | 99.9% | C17H22BrNO3 | 500 MG
SDP
Catalog No. 5000432312
Encompass_Preferred
Galanthamine hydrobromide is a selective, reversible, competitive, alkaloid AChE inhibitor with an IC50 of 0.35 μM. It also acts as a potent allosteric potentiating ligand (APL) for human α3β4, α4β2, and α6β4 nicotinic receptors (nAChRs). This compound is primarily developed for research into Alzheimer's disease (AD).
  • Selective, reversible, competitive alkaloid AChE inhibitor
  • Potent allosteric potentiating ligand of human α3β4, α4β2, α6β4 nicotinic receptors (nAChRs)
  • Developed for Alzheimer's disease (AD) research
  • IC50 of 0.35 μM for AChE
Catalog No. 50-004-32312 Supplier Medchemexpress LLC Supplier No. HYA0009500MG
Galanthamine hydrobromide is a selective, reversible, competitive, alkaloid AChE inhibitor with an IC50 of 0.35 μM. It also acts as a potent allosteric potentiating ligand (APL) for human α3β4, α4β2, and α6β4 nicotinic receptors (nAChRs). This compound is primarily developed for research into Alzheimer's disease (AD).
  • Selective, reversible, competitive alkaloid AChE inhibitor
  • Potent allosteric potentiating ligand of human α3β4, α4β2, α6β4 nicotinic receptors (nAChRs)
  • Developed for Alzheimer's disease (AD) research
  • IC50 of 0.35 μM for AChE