Learn More
Medchemexpress LLC Galanthamine N-Oxide | 134332-50-6 | 1 MG
SDP
Catalog No. 5000419517
Encompass_Preferred
Galanthamine N-Oxide is an alkaloid derived from the bulbs of *Zephyranthes concolor*. This compound acts as an inhibitor of electric eel acetylcholinesterase (AChE) and is recognized for its role in inhibiting substrate accommodation within the active site of *Torpedo californica* AChE (TcAChE), human AChE (hAChE), and human BChE (hBChE) enzymes. It is intended for research use only.
  • Derived from *Zephyranthes concolor* bulbs
  • Inhibits electric eel acetylcholinesterase (AChE)
  • Prominent inhibitor of substrate accommodation in AChE and BChE enzymes
  • Target: Cholinesterase (ChE)
  • Pathway: Neuronal Signaling
  • Appears as a white to off-white solid
Catalog No. 50-004-19517 Supplier Medchemexpress LLC Supplier No. HYN72631MG
Galanthamine N-Oxide is an alkaloid derived from the bulbs of *Zephyranthes concolor*. This compound acts as an inhibitor of electric eel acetylcholinesterase (AChE) and is recognized for its role in inhibiting substrate accommodation within the active site of *Torpedo californica* AChE (TcAChE), human AChE (hAChE), and human BChE (hBChE) enzymes. It is intended for research use only.
  • Derived from *Zephyranthes concolor* bulbs
  • Inhibits electric eel acetylcholinesterase (AChE)
  • Prominent inhibitor of substrate accommodation in AChE and BChE enzymes
  • Target: Cholinesterase (ChE)
  • Pathway: Neuronal Signaling
  • Appears as a white to off-white solid