Learn More
Medchemexpress LLC Galanthamine N-Oxide | 134332-50-6 | 5 MG
SDP
Catalog No. 5000417664
Encompass_Preferred
Galanthamine N-Oxide is an alkaloid obtained from the bulbs of *Zephyranthes concolor*. It functions as an acetylcholinesterase (AChE) inhibitor, exhibiting an EC50 of 26.2 μM against electric eel AChE. This compound is noted as a significant inhibitor of substrate accommodation within the active site of *Torpedo californica* AChE (TcAChE), hAChE, and hBChE enzymes.
  • Alkaloid obtained from the bulbs of Zephyranthes concolor
  • Inhibits electric eel acetylcholinesterase (AChE) with an EC50 of 26.2 μM
  • Significant inhibitor of substrate accommodation in TcAChE, hAChE, and hBChE enzymes
  • Purity of 99.90%
  • Appears as a white to off-white solid
  • Targets cholinesterase (ChE)
  • Involved in neuronal signaling pathways
Catalog No. 50-004-17664 Supplier Medchemexpress LLC Supplier No. HYN72635MG
Galanthamine N-Oxide is an alkaloid obtained from the bulbs of *Zephyranthes concolor*. It functions as an acetylcholinesterase (AChE) inhibitor, exhibiting an EC50 of 26.2 μM against electric eel AChE. This compound is noted as a significant inhibitor of substrate accommodation within the active site of *Torpedo californica* AChE (TcAChE), hAChE, and hBChE enzymes.
  • Alkaloid obtained from the bulbs of Zephyranthes concolor
  • Inhibits electric eel acetylcholinesterase (AChE) with an EC50 of 26.2 μM
  • Significant inhibitor of substrate accommodation in TcAChE, hAChE, and hBChE enzymes
  • Purity of 99.90%
  • Appears as a white to off-white solid
  • Targets cholinesterase (ChE)
  • Involved in neuronal signaling pathways