Learn More
Medchemexpress LLC HY-I0020 5mg Medchemexpress, Galanthaminone CAS:510-77-0 Purity:>98%
SDP
Catalog No. 501933248
Encompass_Preferred
Medchemexpress, HY-I0020 5mg Galanthaminone CAS:510-77-0 Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor; is used for the treatment of mild to moderate Alzheimer's disease and various other memory impairments. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3248 Supplier Medchemexpress LLC Supplier No. HYI00205MG
Medchemexpress, HY-I0020 5mg Galanthaminone CAS:510-77-0 Galanthaminone (Narwedin) is a competitive and reversible cholinesterase (AChE) inhibitor; is used for the treatment of mild to moderate Alzheimer's disease and various other memory impairments. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.