Learn More
Medchemexpress LLC Galaxolide 100g
SDP
Catalog No. 5000206918
Encompass_Preferred
Galaxolide can induce estrogenic activity (Estrogen Receptor/ERR) trigger oxidative stress and genotoxicity and induce EROD and GST (Glutathione S-transferase) enzyme activity[1][2]
Catalog No. 50-002-06918 Supplier Medchemexpress LLC Supplier No. HYW100026100G
Galaxolide can induce estrogenic activity (Estrogen Receptor/ERR) trigger oxidative stress and genotoxicity and induce EROD and GST (Glutathione S-transferase) enzyme activity[1][2]