Learn More
Cayman Chemical Gallic Acid, Purity: ≥98%, CAS: 149-91-7, 50 g
SDP
Catalog No. 502951710
Encompass_Preferred
A phenol with diverse biological activities; scavenges DPPH and hydroxyl radicals in cell-free assays (IC50s = 9.4 and 191 µM, respectively); reverses abscisic acid-induced inhibition of hypocotyl growth in A. caudatus seedlings from 1-100 µM; inhibits morpholine- and sodium nitrite-induced adenocarcinoma formation in mice at 21.8 g/kg; inhibits passive cutaneous anaphylaxis in mice at 50 mg/kg
Catalog No. 50-295-1710 Supplier Cayman Chemical Supplier No. 1184650 G
A phenol with diverse biological activities; scavenges DPPH and hydroxyl radicals in cell-free assays (IC50s = 9.4 and 191 µM, respectively); reverses abscisic acid-induced inhibition of hypocotyl growth in A. caudatus seedlings from 1-100 µM; inhibits morpholine- and sodium nitrite-induced adenocarcinoma formation in mice at 21.8 g/kg; inhibits passive cutaneous anaphylaxis in mice at 50 mg/kg