Learn More
Santa Cruz Biotechnology GALCTIN3 ANTBDY(B2C10)200UG/ML
Supplier:  Santa Cruz Biotechnology SC32790
Encompass
galectin-3 Antibody (B2C10): sc-32790 200 µg/ml. •Anti-galectin-3 Antibody (B2C10) is a mouse monoclonal IgG1 (kappa light chain) galectin-3 antibody provided at 200 µg/ml•raised against recombinant Galectin-3 of human origin with an epitope mapping within the first 18 amino acids•Anti-galectin-3 Antibody (B2C10) is recommended for detection of galectin-3 of mouse, rat and human origin by WB, IP, IF, IHC(P) and FCM
Catalog No. NC1877138