Learn More
Aviva Systems Biology Corp GALE Antibody - middle region (ARP54394_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54394_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALE. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GALE. Alternate names: SDR1E1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-285-920