Learn More
ACROBiosystems ACROBiosystems Human Galectin-3 Protein, His Tag
Supplier:  ACROBiosystems GA3H5129200UG
Encompass_Preferred
Recombinant Protein;200UG;Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contains AA Met 1 - Ile 250 (Accession # NP_002297).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6969