Learn More
BioVendor, LLC Galectin-3 Human Hs Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL011R
Encompass
Galectin-3 is a member of the galectin family, which are ?-galactoside-binding lectins with evolutionary conserved carbohydrate-recognition domain. The protein is expressed by various immune cells and is readily secreted by injured and inflammatory cells. Galectin-3 is also a novel candidate biomarker for the diagnosis, analysis, and prognosis of various cardiac diseases, including heart failure. Gallectin-3 has also been linked to kidney disease in clinical studies, and its inhibition appears to improve renal disease. One kit (96 wells). Type: sandwich ELISA. Sample type: serum, plasma. Sample requirement: 7 ¿l/well. Calibration range: 125-4000pg/ml. Limit of detection: 30pg/ml. Other names: Galactose specific lectin 3, Mac-2 antigen, 35 kDa lectin, Carbohydrate binding protein 35, Laminin binding protein.
Catalog No. 50-240-6374