Learn More
Medchemexpress LLC Galectin-3/LGALS3 M 100ug | 100ug
SDP
Catalog No. 5000194764
Encompass_Preferred
Galectin-3 (LGALS3) is involved in multiple activities including IgE binding and signaling receptor binding It negatively regulates T-cell receptor signaling and modulates immune responses by modulating endocytosis and lymphocyte activation Galectin-3/LGALS3 Protein Mouse is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by E coli with tag free
Catalog No. 50-001-94764 Supplier Medchemexpress LLC Supplier No. HYP79079100UG
Galectin-3 (LGALS3) is involved in multiple activities including IgE binding and signaling receptor binding It negatively regulates T-cell receptor signaling and modulates immune responses by modulating endocytosis and lymphocyte activation Galectin-3/LGALS3 Protein Mouse is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by E coli with tag free