Learn More
Medchemexpress LLC Galectin-3/LGALS3 M 10ug | 10ug
SDP
Catalog No. 5000187053
Encompass_Preferred
Galectin-3/LGALS3 protein binds IgE and cooperates with integrins -3 and -1 to promote endothelial cell migration It contributes to epithelial cell differentiation and acts as a splicing factor in the nucleus Galectin-3/LGALS3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87053 Supplier Medchemexpress LLC Supplier No. HYP7768410UG
Galectin-3/LGALS3 protein binds IgE and cooperates with integrins -3 and -1 to promote endothelial cell migration It contributes to epithelial cell differentiation and acts as a splicing factor in the nucleus Galectin-3/LGALS3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by HEK293 with C-His labeled tag