Learn More
ABclonal Technology Galectin 3/LGALS3 Rabbit mAb
SDP
Catalog No. 5000006739
Encompass_Preferred
This gene encodes a member of the galectin family of carbohydrate binding proteins Members of this protein family have an affinity for beta-galactosides The encoded protein is characterized by an N-terminal proline-rich tandem repeat domain and a single C-terminal carbohydrate recognition domain This protein can self-associate through the N-terminal domain allowing it to bind to multivalent saccharide ligands This protein localizes to the extracellular matrix the cytoplasm and the nucleus This protein plays a role in numerous cellular functions including apoptosis innate immunity cell adhesion and T-cell regulation The protein exhibits antimicrobial activity against bacteria and fungi Alternate splicing results in multiple transcript variants
Catalog No. 50-000-06739 Supplier ABclonal Technology Supplier No. A2276820UL
This gene encodes a member of the galectin family of carbohydrate binding proteins Members of this protein family have an affinity for beta-galactosides The encoded protein is characterized by an N-terminal proline-rich tandem repeat domain and a single C-terminal carbohydrate recognition domain This protein can self-associate through the N-terminal domain allowing it to bind to multivalent saccharide ligands This protein localizes to the extracellular matrix the cytoplasm and the nucleus This protein plays a role in numerous cellular functions including apoptosis innate immunity cell adhesion and T-cell regulation The protein exhibits antimicrobial activity against bacteria and fungi Alternate splicing results in multiple transcript variants