Learn More
Enzo Life Sciences Galectin-3 monoclonal antibody (A3A12) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804284C100
Encompass_Preferred
Clone: A3A12. Isotype: IgG1. Immunogen: Recombinant human galectin-3. Species reactivity: Human, Mouse, Rat. Applications: ELISA, Flow Cytometry, IHC (PS), WB. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9854