Learn More
Sino Biological Human Galectin 3 / LGALS3 Protein (His Tag) 100ug
Supplier:  Sino Biological 10289H08H1100
Encompass_Preferred
A DNA sequence encoding the human LGALS3 (NP_002297.2) (Ala2-Ile250) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6451