Learn More
Medchemexpress LLC Galectin-4 Mouse H 100ug | 100ug
SDP
Catalog No. 5000194197
Encompass_Preferred
Galectin-4 is a lactose-binding protein belonging to the galectin family that exhibits specific affinities for a range of structurally related sugars This multifunctional protein is noteworthy for its functionality as a monomer highlighting its individual molecular structure and suggesting that it may exert its biological effects through independent interactions rather than as part of a larger complex Galectin-4 Protein Mouse (HEK293 His) is the recombinant mouse-derived Galectin-4 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-94197 Supplier Medchemexpress LLC Supplier No. HYP78694100UG
Add to Cart
Add to Cart
Galectin-4 is a lactose-binding protein belonging to the galectin family that exhibits specific affinities for a range of structurally related sugars This multifunctional protein is noteworthy for its functionality as a monomer highlighting its individual molecular structure and suggesting that it may exert its biological effects through independent interactions rather than as part of a larger complex Galectin-4 Protein Mouse (HEK293 His) is the recombinant mouse-derived Galectin-4 protein expressed by HEK293 with N-His labeled tag