Learn More
ACROBiosystems ACROBiosystems Human Galectin-9 / LGALS9 Protein, His Tag
Supplier:  ACROBiosystems LG9H52441MG
Encompass_Preferred
Recombinant Protein;1MG;Human Galectin-9, His Tag (LG9-H5244) is expressed from human 293 cells (HEK293). It contains AA Ala 2 - Thr 323 (Accession # BAA31542).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.6 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7326