Learn More
ACROBiosystems Recombinant Protein;Human Galectin-3 Protein;E.coli;1MG;GA3,Galectin-3,LGALS3,MAC2,Gal-3,35 kDa lectin,CBP 35,GALBP,IgE-binding protein,L-31,Laminin-binding protein,Lectin L-29,Mac-2 antigen,Galectin 3,LGALS-3,MAC-2,Gal3,L31,Galectin3
Catalog No. 501658269
Encompass_Preferred
Recombinant Protein;Human Galectin-3 Protein;1MG;Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contains AA Met 1 - Ile 250 (Accession # NP_002297).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8269 Supplier ACROBiosystems Supplier No. GA3H51291MG
Recombinant Protein;Human Galectin-3 Protein;1MG;Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contains AA Met 1 - Ile 250 (Accession # NP_002297).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE).