Learn More
Cayman Chemical GalectIn9ShortVarInthum50ug
SDP
Catalog No. 502947788
Encompass_Preferred
Source: Recombinant N-terminal human IgG1 Fc-tagged galectin 9 expressed in HEK293 cells • Amino acids: 2-323 • MW: 61.8 kDa
Catalog No. 50-294-7788 Supplier Cayman Chemical Supplier No. 3201250 UG
Source: Recombinant N-terminal human IgG1 Fc-tagged galectin 9 expressed in HEK293 cells • Amino acids: 2-323 • MW: 61.8 kDa