Learn More
Medchemexpress LLC Galegine hemisulfate | 14279-86-8 | 176.23 | 5 MG
SDP
Catalog No. 5000417150
Encompass_Preferred
Galegine hemisulfate is a guanidine derivative that contributes to weight loss in mice. It activates AMPK in 3T3-L1 adipocytes, L6 myotubes, H4IIE rat hepatoma cells, and HEK293 human kidney cell lines. It also exhibits antibacterial activity with a minimum inhibitory concentration of 4 mg/L against *Staphylococcus aureus* strains.
  • Contributes to weight loss in mice
  • Activates AMPK in various cell lines (3T3-L1 adipocytes, L6 myotubes, H4IIE rat hepatoma, HEK293 human kidney cells)
  • Has antibacterial activity against *Staphylococcus aureus* strains
  • Stimulates insulin-independent glucose uptake by 3T3-L1 adipocytes and L6 myotubes in a concentration-dependent manner
  • Reduces basal glycerol release and markedly reduces Isoprenaline-stimulated glycerol release from 3T3-L1 adipocytes
Catalog No. 50-004-17150 Supplier Medchemexpress LLC Supplier No. HYN0930A5MG
Galegine hemisulfate is a guanidine derivative that contributes to weight loss in mice. It activates AMPK in 3T3-L1 adipocytes, L6 myotubes, H4IIE rat hepatoma cells, and HEK293 human kidney cell lines. It also exhibits antibacterial activity with a minimum inhibitory concentration of 4 mg/L against *Staphylococcus aureus* strains.
  • Contributes to weight loss in mice
  • Activates AMPK in various cell lines (3T3-L1 adipocytes, L6 myotubes, H4IIE rat hepatoma, HEK293 human kidney cells)
  • Has antibacterial activity against *Staphylococcus aureus* strains
  • Stimulates insulin-independent glucose uptake by 3T3-L1 adipocytes and L6 myotubes in a concentration-dependent manner
  • Reduces basal glycerol release and markedly reduces Isoprenaline-stimulated glycerol release from 3T3-L1 adipocytes