Learn More
Medchemexpress LLC Galegine hemisulfate | 14279-86-8 | MFCD16293919 | 99.7% | 176.23 g/mol | C6H13N3·1/2H2O·4S | 10 MG
SDP
Catalog No. 5000296072
Encompass_Preferred
Galegine hemisulfate is a guanidine derivative used in research to activate AMP-activated protein kinase (AMPK), stimulate insulin-independent glucose uptake, and investigate antibacterial effects. It is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Activates AMPK in adipocytes, myotubes, and kidney cell lines.
  • Stimulates insulin-independent glucose uptake in adipocytes and myotubes.
  • Exhibits antibacterial activity against Staphylococcus aureus (MIC 4 mg/L).
  • Reduces body weight in mice in dietary administration studies.
  • Provided as a solid with high purity and specified storage conditions.
Catalog No. 50-002-96072 Supplier Medchemexpress LLC Supplier No. HYN0930A10MG
Galegine hemisulfate is a guanidine derivative used in research to activate AMP-activated protein kinase (AMPK), stimulate insulin-independent glucose uptake, and investigate antibacterial effects. It is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Activates AMPK in adipocytes, myotubes, and kidney cell lines.
  • Stimulates insulin-independent glucose uptake in adipocytes and myotubes.
  • Exhibits antibacterial activity against Staphylococcus aureus (MIC 4 mg/L).
  • Reduces body weight in mice in dietary administration studies.
  • Provided as a solid with high purity and specified storage conditions.