Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galena, naturally occurring mineral, grains, approximately 0.06-0.19in

Catalog No. p-4904540
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4264003 1 g
AA4264006 5 g
AA4264014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4264003 Supplier Thermo Scientific Chemicals Supplier No. 04264003

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 1314-87-0 | PbS | 239.30 g/mol

Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name sulfanylidenelead
SMILES [S--].[Pb++]

Specifications

Color Black
Physical Form Powder, Flake, Crystalline, Beads, Lump, Etc.
Quantity 1 g
Solubility Information Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide.
Formula Weight 239.25
Odor Odorless
Chemical Name or Material Galena, naturally occurring mineral, grains, approximately 0.06-0.19 in.
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-246-6
RTECSNumber OG4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.