Learn More
Cayman Chemical Gallic Acid-d2, Purity: ≥99% deutereated forms (d1-d2), CAS: 294660-92-7, 100 mg
SDP
Catalog No. 502910653
Encompass_Preferred
An internal standard for the quantification of gallic acid by GC- or LC-MS
Catalog No. 50-291-0653 Supplier Cayman Chemical Supplier No. 35132100 MG
An internal standard for the quantification of gallic acid by GC- or LC-MS