Learn More
Medchemexpress LLC Galicaftor | 1918143-53-9 | 98.6% | 559.46 g/mol | C28H21F4NO7 | 5 MG
SDP
Catalog No. 5000263356
Encompass_Preferred
Galicaftor is a small-molecule, orally active CFTR corrector used in cystic fibrosis research. It is provided for laboratory research use only and is available as a solid and as DMSO solutions; analytical data and storage recommendations are supplied by the manufacturer.
  • Potent CFTR corrector activity suitable for preclinical studies.
  • High purity for reliable analytical and biological assays.
  • Available as solid packs and ready-to-use DMSO solutions.
  • Defined storage conditions to maintain stability.
  • Chemical identification data (CAS, formula, molecular weight) provided.
Catalog No. 50-002-63356 Supplier Medchemexpress LLC Supplier No. HY1111115MG
Galicaftor is a small-molecule, orally active CFTR corrector used in cystic fibrosis research. It is provided for laboratory research use only and is available as a solid and as DMSO solutions; analytical data and storage recommendations are supplied by the manufacturer.
  • Potent CFTR corrector activity suitable for preclinical studies.
  • High purity for reliable analytical and biological assays.
  • Available as solid packs and ready-to-use DMSO solutions.
  • Defined storage conditions to maintain stability.
  • Chemical identification data (CAS, formula, molecular weight) provided.