Learn More
Medchemexpress LLC Galidesivir hydrochloride | 222631-44-9 | 99.9% | 301.73 | 25 MG
SDP
Catalog No. 5000384070
Encompass_Preferred
Galidesivir hydrochloride is an adenosine analog and a direct-acting antiviral agent that disrupts viral RNA-dependent RNA polymerase (RdRp) activity. It is active in vitro against many RNA viral pathogens, including filoviruses and emerging infectious agents like MERS-CoV, SARS-CoV, and SARS-CoV-2. It inhibits some negative-sense RNA viruses with EC50s ranging from ~3 to ~68 μM.
  • Adenosine analog
  • Direct-acting antiviral agent
  • Disrupts viral RNA-dependent RNA polymerase (RdRp) activity
  • Active against many RNA viral pathogens
  • Inhibits negative-sense RNA viruses
Catalog No. 50-003-84070 Supplier Medchemexpress LLC Supplier No. 25MGHY18649
Galidesivir hydrochloride is an adenosine analog and a direct-acting antiviral agent that disrupts viral RNA-dependent RNA polymerase (RdRp) activity. It is active in vitro against many RNA viral pathogens, including filoviruses and emerging infectious agents like MERS-CoV, SARS-CoV, and SARS-CoV-2. It inhibits some negative-sense RNA viruses with EC50s ranging from ~3 to ~68 μM.
  • Adenosine analog
  • Direct-acting antiviral agent
  • Disrupts viral RNA-dependent RNA polymerase (RdRp) activity
  • Active against many RNA viral pathogens
  • Inhibits negative-sense RNA viruses