Learn More
Enzo Life Sciences Galiellalactone (0.5mg). CAS: 133613-71-5
SDP
Supplier:  Enzo Life Sciences ALX350336MC05
Encompass_Preferred
Originally isolated from Galiella rufa as a plant growth regulator. Inhibits IL-6 mediated signal transduction by blocking the binding of the activated STAT3 dimers to their DNA binding sites without inhibiting the tyrosine and serine phosphorylation site of the STAT3 transcription factor. Formulation: Lyophilized. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8849