Learn More
Fine Science Tools Galilean Loupes, 3.5x, 300 mm Working Distance, 77 g
SDP
Catalog No. NC1310573
Encompass
These loupes are lightweight and easy to use. Made from the finest optical coated lenses which deliver excellent clarity and high resolution. The magnifier loupes are mounted onto a strong yet flexible wraparound safety frame with a headband for stability and comfort. Ability to flip loupes up for use as safety glasses. Comes with mini screwdriver for fine adjustments, micro cloth, and storage case.Magnification: 3.5x; Working Distance: 300mm (16"): Weight: 77g
Catalog No. NC1310573 Supplier Fine Science Tools Supplier No. 2805035
These loupes are lightweight and easy to use. Made from the finest optical coated lenses which deliver excellent clarity and high resolution. The magnifier loupes are mounted onto a strong yet flexible wraparound safety frame with a headband for stability and comfort. Ability to flip loupes up for use as safety glasses. Comes with mini screwdriver for fine adjustments, micro cloth, and storage case.Magnification: 3.5x; Working Distance: 300mm (16"): Weight: 77g