Learn More
ABclonal Technology GALK1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15274500UL
Encompass_Preferred
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts during infancy and presenile cataracts in the adult population.
Catalog No. 50-001-06787