Learn More
TARGETMOL CHEMICALS INC GALLEIN 100MG
SDP
Catalog No. 502577017
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gallein (Mordant violet 25) is a small molecule inhibitor of G(beta)(gamma) activity. purity: 95%
Catalog No. 50-257-7017 Supplier TARGETMOL CHEMICALS INC Supplier No. T3040100MG
Also available in 5 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gallein (Mordant violet 25) is a small molecule inhibitor of G(beta)(gamma) activity. purity: 95%