Learn More
Medchemexpress LLC HY-D0254 10mg , Gallein CAS:2103-64-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD025410MG
Encompass_Preferred
HY-D0254 10mg Gallein CAS: 2103-64-2 Gallein is a G protein βγ (Gβγ) subunit signalling inhibitor. Gallein disrupts the interaction of Gβγ subunits with the PI3Kγ. Anticancer agent. Gallein used as a red dye, an acid-base indicator and as a detection reagent for phosphates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1156