Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallein 85.0+%, TCI America™
SDP

Catalog No. A060125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A060125G 25 g
A06011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A060125G Supplier TCI America Supplier No. A060125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2103-64-2
Molecular Formula C20H12O7
Molecular Weight (g/mol) 364.309
MDL Number MFCD00041839
InChI Key PHLYOKFVXIVOJC-UHFFFAOYSA-N
Synonym gallein, alizarin violet, pyrogallolphthalein, c.i. mordant violet 25, 4,5-dihydroxyfluorescein, mordant violet 25, 3',4',5',6'-tetrahydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, alizarine violet, unii-8l0084u2qr, fluorescein, 4',5'-dihydroxy
PubChem CID 73685
IUPAC Name 3',4',5',6'-tetrahydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)O)OC5=C3C=CC(=C5O)O

Specifications

Color Black
Quantity 25 g
Formula Weight 364.31
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gallein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.