Learn More
Medchemexpress LLC HY-N0523 100mg Medchemexpress, Gallic acid CAS:149-91-7 Purity:>98%
SDP
Catalog No. 501933606
Encompass_Preferred
Medchemexpress, HY-N0523 100mg Gallic acid CAS:149-91-7 Gallic acid is an antioxidant which can inhibit both COX-2 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3606 Supplier Medchemexpress LLC Supplier No. HYN0523100MG
Medchemexpress, HY-N0523 100mg Gallic acid CAS:149-91-7 Gallic acid is an antioxidant which can inhibit both COX-2 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.